Anti-HSPB1 (pSer82) Rabbit Monoclonal Antibody [clone: CB2] (APC)
Catalog # ABNOMAB23516.100
Supplier: Abnova
Specifications
- Antigen symbol:HSPB1
- Clonality:Monoclonal
- Clone:CB2
- Conjugation:APC (Allophycocyanin)
- Host:Rabbit
- Reactivity:Human
- Modification:pSer82
- Immunogen:Rabbit monoclonal antibody raised against synthetic phosphopeptide of human HSPB1.
- Pk:100 AU