Order Entry
Spain
ContactUsLinkComponent
96 Well Plate, RCH TRACE-N
96 Well Plate, RCH TRACE-N
Catalog # TCAT610-0101RCH
Supplier:  Tecan
96 Well Plate, RCH TRACE-N
Catalog # TCAT610-0101RCH
Supplier:  Tecan
Specifications
  • Description:
    Plate RCH TRACE-N; 1 ml, 96, 10 mg
  • Pk:
    1
Specifications
About this item
End-capped C18 chemistry.
  • Reversed-phase
  • Liquid handler compatible
A 10 µm micro particulate with end-capped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.