Order Entry
Spain
ContactUsLinkComponent
SPE Column, NBE PSCX
SPE Column, NBE PSCX
Catálogo # TCAT687-0051R-NBE
Supplier:  Tecan
SPE Column, NBE PSCX
Catálogo # TCAT687-0051R-NBE
Supplier:  Tecan

Specifications

  • Particle Size:
    15 µm
  • Pore size:
    200 Å
  • Capacity:
    1 ml
  • Description:
    Column NBE PSCX
  • Bed weight:
    5 mg
  • Phase:
    PSCX
  • Pk:
    96

Specifications

About this item

Polymer micro particulate with strong cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample preparation. Ideal for basic target compounds.

  • Ion-exchange
  • Liquid handler compatible