Order Entry
Spain
ContactUsLinkComponent
Cerex® OFXQ SPE Column
Cerex® OFXQ SPE Column
Catalog # TCAT635-0101R
Supplier:  Tecan
Cerex® OFXQ SPE Column
Catalog # TCAT635-0101R
Supplier:  Tecan

Specifications

  • Particle Size:
    30 µm
  • Capacity:
    1 ml
  • Description:
    Column Cerex OFXQ
  • Bed weight:
    10 mg
  • Phase:
    OFXQ
  • Pk:
    96

Specifications

About this item

Proprietary blend of ion-exchange and water-wettable polymer targeted towards oral fluid collection devices.

  • Ion-exchange
  • Liquid handler compatible

A proprietary polymer resin blend of ion exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.