96 Well Plate RCH CEREX HPWCX
Supplier: Tecan
Certificates
About this item
A 25 µm polymer micro particulate with weak cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample preparation.
- Cerex
- Ion-Exchange
- Liquid handler compatible
Frequently Bought Together