Order Entry
Spain
ContactUsLinkComponent
96 Well Plate, RCH CEREX MAESTRO
96 Well Plate, RCH CEREX MAESTRO
Catalog # TCAT613-0151RCH
Supplier:  Tecan
96 Well Plate, RCH CEREX MAESTRO
Catalog # TCAT613-0151RCH
Supplier:  Tecan
Specifications
  • No. of wells:
    96
  • Description:
    96-well plate for RCH CEREX MAESTRO 1 ml
  • Phase:
    MAESTRO
  • Filling:
    15 mg
  • Pk:
    1
Specifications
About this item
Silica-based sorbent with C18 bonding in proprietary coverage.
  • Reversed-phased
  • Liquid handler compatible
Uses a combination of filtration and sorbent interaction. Ideal for pass-through sample preparation techniques to remove phospholipids, proteins and particulates from sample matrices.