Order Entry
Spain
ContactUsLinkComponent
CM Sephadex™ C-25 Ion Exchange Chromatography Media
CM Sephadex™ C-25 Ion Exchange Chromatography Media
Supplier:  Cytiva
Certificates
undefined
CM Sephadex™ C-25 Ion Exchange Chromatography Media
Supplier:  Cytiva

Specifications

  • pH range:
    6 - 10
  • Matrix:
    Cross-linked dextran
  • Ligand:
    Carboxymethyl
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    lgG (Mr 160 000)- 1,6 mg/ml drained medium), Bovine COHb (Mr 69 000)- 70 mg/ml drained medium), Ribonuclease (Mr 13 700)- 190 mg/ml drained medium)
  • Total ionic capacity:
    4 - 5 mmol/g
  • Description:
    CM Sephadex™ C-25

Specifications

Frequently Bought Together