Order Entry
Spain
ContactUsLinkComponent
Culti-Loops™ Fusobacterium nucleatum subsp. nucleatum ATCC™ 25586™
Culti-Loops™ Fusobacterium nucleatum subsp. nucleatum ATCC™ 25586™
Catalog # OXOIR4602010
CAS Number:  
undefined
Culti-Loops™ Fusobacterium nucleatum subsp. nucleatum ATCC™ 25586™
Catalog # OXOIR4602010
CAS Number:  
Specifications
  • Pk:
    5
  • Description:
    Fusobacterium nucleatum subsp. nucleatum ATCC™ 25586™
Specifications
About this item
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, or streaked directly onto an appropriate media.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.