Order Entry
Spain
ContactUsLinkComponent
Culti-Loops™ Enterococcus faecium ATCC™ 19434™
Culti-Loops™ Enterococcus faecium ATCC™ 19434™
Catalog # OXOIR4609045
CAS Number:  
undefined
Culti-Loops™ Enterococcus faecium ATCC™ 19434™
Catalog # OXOIR4609045
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Pk:
    5
  • Description:
    Enterococcus faecium ATCC™ 19434™
Specifications
About this item
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, or streaked directly onto an appropriate media.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.