Locust bean gum FCC
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:9000-40-2
:
MDL Number:MFCD00131257
Storage Temperature:Ambient
Specification Test Results
- Galactomannans:0.75
- Acid-Insoluble Matter:max.4.0 %
- Arsenic (As):max. 3 mg/kg
- Ash (Total):max. 1.2 %
- Lead (Pb):max. 5 mg/kg
Specification Test Results
Frequently Bought Together