Order Entry
Spain
ContactUsLinkComponent
Hectorite
Hectorite
Catalog # SPCMH1406-500GM
Supplier:  Spectrum Chemical
CAS Number:  
Hectorite
Catalog # SPCMH1406-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly pail
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :GHS08
Storage Temperature:Ambient
CAS Number:12173-47-6
Formula:Na₀.₆₇(Mg,Li)₆Si₈O₂₀(OH,F)₄