Acid red 33
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:3567-66-6
:
Formula:C₁₆H₁₁N₃Na₂O₇S₂
MDL Number:MFCD00071877
:467.378 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together
Methanol : Water (30:70) % (v/v), LiChrosolv®, Supelco®
Supplier: Merck
p-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran 99.8% unstabilised for HPLC
Supplier: Thermo Fisher Scientific
Hydrindantin dihydrate, Supelco®
Supplier: Merck
Sodium hexanitritocobaltate(III)
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide 99.5+% for analysis
Supplier: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Water, pure, demineralised
Supplier: Thermo Fisher Scientific
Bromoform 96% stabilised
Supplier: Thermo Fisher Scientific
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
p-Xylene ≥99.0% (by GC)
Supplier: TCI
Cyclopentane for HPLC
Supplier: Thermo Fisher Scientific
Chloroform stabilised for HPLC
Supplier: Thermo Fisher Scientific
Pyridine 99+%, extra pure for non-aqueous titration
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
Supplier: Honeywell Chemicals