Order Entry
Spain
ContactUsLinkComponent
Cyclohexylamine hydrochloride
Cyclohexylamine hydrochloride
Catalog # SPCMC2127-25KGBL
Supplier:  Spectrum Chemical
CAS Number:  
Cyclohexylamine hydrochloride
Catalog # SPCMC2127-25KGBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Fiber drum
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :135.64 g/mol
Storage Temperature:Ambient
CAS Number:4998-76-9
Formula:C₆H₁₃N
MDL Number:MFCD00050540