1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione
Catalog # SPCMA2682-25GM
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Glass, amber
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Slightly Yellow-
- Assay:97.0 % min.
- Melting Point:81 - 86°C
Specification Test Results
About this item
:
:310.39 g/mol
Storage Temperature:Ambient
CAS Number:70356-09-1
Formula:C₂₀H₂₂O₃
MDL Number:MFCD00210252