Lactobionic acid ≥98.0% Ph. Eur.
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:96-82-2
:
Formula:C₁₂H₂₂O₁₂
MDL Number:MFCD00078147
:358.30 g/mol
Storage Temperature:Ambient
Specification Test Results
- Assay (Anhydrous Basis):98.0 - 102.0%
- Appearance of Solution:To Pass Test
- Specific Optical Rotation:+23.0 to +29.0
- Reducing Sugars (As Glucose):max. 0.2 %
- Heavy Metals:max. 20 ppm
Specification Test Results
Frequently Bought Together