Dextran, M.W. ~ 40,000, powder
Catalog # SPCMDE124-1KG
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
Storage Temperature:Ambient
EINECS:232-677-5
CAS Number:9004-54-0
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00130935