Order Entry
Spain
ContactUsLinkComponent
Magnesium hydroxide ≥95%, powder USP
Magnesium hydroxide ≥95%, powder USP
  SPCMMA115-2.5KG
 :  
Magnesium hydroxide ≥95%, powder USP
  SPCMMA115-2.5KG
 :  

 

  • Pack type:
    Poly pail
  • Pk:
    2,5 kg
  • Seq:
    4076

 

 

  • Assay:
    95% Min.

 

 

 :
 :58.32 g/mol
 :13,05693
 :2,3261…2,3631 g/cm³ (20 °C)
 :Ambient
 :215-170-3
 :1309-42-8
 :Mg(OH)₂
 :MFCD00011104