Order Entry
Spain
ContactUsLinkComponent
Dibutyl sebacate ≥98%
Dibutyl sebacate ≥98%
Catalog # SPCMD2915-1LT
Supplier:  Spectrum Chemical
CAS Number:  
Dibutyl sebacate ≥98%
Catalog # SPCMD2915-1LT
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    98% Min.

Specification Test Results

About this item

 :
 :314.47 g/mol
Boiling Pt:180 °C (1013 hPa)
Melting Pt:–11 °C
Density:0,936
Storage Temperature:Ambient
EINECS:203-672-5
 :GHS07
CAS Number:109-43-3
Formula:C₁₈H₃₄O₄
MDL Number:MFCD00027218