Gum arabic, spray-dried powder FCC
Supplier: Spectrum Chemical
New Product
Certificates
About this item
CAS Number:9000-01-5
:
MDL Number:MFCD00081264
Storage Temperature:Ambient
Specification Test Results
- Loss On Drying:max. 15.0%
- Arsenic (As):max.3 mg/kg
- Acid Insoluble Ash:max. 0.5 %
- Total Ash:max. 4.0 %
- Insoluble Matter:max. 1.0 %
Specification Test Results
Frequently Bought Together