Order Entry
Spain
ContactUsLinkComponent
L(-)-Glutathione (reduced form) ≥96%, reduced
L(-)-Glutathione (reduced form) ≥96%, reduced
  SPCMGL146-25GM
 :  
L(-)-Glutathione (reduced form) ≥96%, reduced
  SPCMGL146-25GM
 :  

 

  • Pack type:
    Glass, amber
  • Pk:
    25 mg
  • Seq:
    2315

 

 

  • Assay:
    96% Min.

 

 

 :
 :307.33 g/mol
 :13,04488
 :185…195 °C
 :1,475 g/cm³ (20 °C)
 :Refrigerator
 :200-725-4
 :70-18-8
 :C₁₀H₁₇N₃O₆S
 :MFCD00065939