Barium peroxide, Reagent Grade
Catalog # SPCMB1053-12KGBL
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pk:12 kg
- Seq:4187
Specifications
About this item
:
:169.33 g/mol
Storage Temperature:Ambient
UN:1449
ADR:5.161,II
CAS Number:1304-29-6
Formula:BaO₂
MDL Number:MFCD00003454