Order Entry
Spain
ContactUsLinkComponent
Acetic acid glacial FCC
Acetic acid glacial FCC
Catalog # SPCMA1009-4LTPV
Supplier:  Spectrum Chemical
CAS Number:  
undefined
Acetic acid glacial FCC
Catalog # SPCMA1009-4LTPV
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, poly-coated
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Assay:
    99.5 - 100.5%
  • Solidification Point:
    15.6°C min.
  • Nonvolatile Residue:
    max. 0.005 %
  • Lead (Pb):
    max. 0.5 mg/kg
  • Readily Oxidizable Substances:
    To Pass Test

Specification Test Results

About this item

 :
 :60.05 g/mol
Density:1,049
Storage Temperature:Ambient
UN:2789
ADR:8.3,II
 :GHS02
 :GHS07
 :GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152