Anti-C1orf33 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-15064R-CY3
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:C1orf33
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:1-80/239
- Form:Liquid
- Antigen synonyms:Ribosomal protein large P0 like|MRTO4|Ribosomal protein P0 like protein|MRT4_HUMAN|MRT4 mRNA turnover 4 homolog|MRT 4|dJ657E11.4|MRT4|MRTO 4|mRNA turnover 4 homolog|60S acidic ribosomal protein PO
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:28 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C1orf33
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
C1orf33
Type: Primary
Antigen: C1orf33
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 1-80/239
Host: Rabbit
Isotype: IgG
Reactivity: