- Antibody type:Primary
- Antigen name:NNP1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:261-360/461
- Form:Liquid
- Gene ID:8568
- Antigen synonyms:NNP 1|Protein NNP 1|Novel nuclear protein 1|RRP1-like protein|RRP1 like protein|RRP1A|Ribosomal RNA processing 1 homolog|Nnp1 homolog nucleolar protein|Nucleolar protein Nop52|Protein NNP1|rrp1|NNP-1|RRP1_HUMAN|NNP1|Ribosomal RNA processing protein 1 homolog A
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human NNP1/RRP1 like protein
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Novel nuclear protein 1 (NNP-1), also known as RRP1-like protein or Nucleolar protein Nop52, is a 461 amino acid protein belonging to the RRP1 family. Localised to the nucleolus, NNP-1 has simian virus 40-type and bipartite nuclear localisation signals and four coiled-coil domains within its C-terminal region. NNP-1 has been found to play an important role in the generation of 28S rRNA in the late processing steps of ribosome biogenesis. At the end of mitosis, nucleolar proteins assemble in a sequential order during the rebuilding of the nucleolus. NNP-1 assembles after Fibrillarin and C23, and simultaneously with B23 and POP1 in the prenucleolar body pathway.
Type: Primary
Antigen: NNP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 261-360/461
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat