Anti-Capicua/CIC Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-11721R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Capicua/CIC
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Environmentally Preferable:
- Epitope:1475-1560/1608
- Form:Liquid
- Gene ID:23152
- Antigen synonyms:CIC|KIAA0306|Protein capicua homolog
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:164 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Capicua
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Transcriptional repressor which may play a role in development of the central nervous system (CNS).
Type: Primary
Antigen: Capicua/CIC
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1475-1560/1608
Host: Rabbit
Isotype: IgG
Reactivity: Mouse