Anti-Syntrophin-3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-10385R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Syntrophin-3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:131-230/540
- Form:Liquid
- Gene ID:6645
- Antigen synonyms:SNTB2_HUMAN|D16S2531E|SNT2B2|Beta-2-syntrophin|59 kDa dystrophin-associated protein A1 basic component 2|SNTB2|EST25263|Syntrophin 3|SNTL|SNT3|Syntrophin-like
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:59 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Syntrophin-3
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Dystrophin is a large, rod-like cytoskeletal protein found at the inner surface of muscle fibers. Dystrophin is missing in Duchenne Muscular Dystrophy patients and is present in reduced amounts in Becker Muscular Dystrophy patients. The protein encoded by this gene is a peripheral membrane protein found associated with dystrophin and dystrophin-related proteins. This gene is a member of the syntrophin gene family, which contains at least two other structurally-related genes.
Type: Primary
Antigen: Syntrophin-3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 131-230/540
Host: Rabbit
Isotype: IgG
Reactivity: