Order Entry
Spain
ContactUsLinkComponent
Anti-SIGLEC9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-SIGLEC9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-9874R-A680
Supplier:  Bioss
Anti-SIGLEC9 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-9874R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SIGLEC9
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    166-260/463
  • Form:
    Liquid
  • Gene ID:
    27180
  • Antigen synonyms:
    SIGL9_HUMAN|SIGLEC9|Siglec-9|OB binding protein like|CDw329|sialic acid binding Ig like lectin 9|OBBP LIKE|Sialic acid-binding immunoglobulin like lectin 9|Siglec 9|Protein FOAP 9|Protein FOAP-9|CD329|Sialic acid-binding Ig-like lectin 9
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human SIGLEC9
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Two families of mammalian lectin-like adhesion molecules bind glycoconjugate ligands in a sialic acid-dependent manner: the selectins and the sialoadhesins. The sialic acid-binding immunoglobulin superfamily lectins, designated siglecs or sialoadhesins, are immunoglobulin superfamily members recognizing sialylated ligands. The common sialic acids of mammalian cells are N-acetyl-neuraminic acid (Neu5Ac) and N-glycolyl-neuraminic acid (Neu5Gc). Siglec-1 mediates local cell-cell interactions in lymphoid tissues and can be detected at contact points of macrophages with other macrophages, sinus-lining cells and reticulum cells. Siglec-7, highly expressed in monocytes and resident blood cells, but not in parenchymatous cells, mediates inhibition of natural killer cell cytotoxicity. Siglec-9 is closely homologous to Siglec-7; the gene encoding it maps to chromosome 19q13.41 in humans. It is highly expressed in peripheral blood leukocytes (but not eosinophils), liver, bone marrow, placenta and spleen. Siglec-8, a type I membrane protein, is selectively expressed on human eosinophils, basophils and mast cells, where it regulates their function and survival.

Type: Primary
Antigen: SIGLEC9
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 166-260/463
Host: Rabbit
Isotype: IgG
Reactivity: Human