Anti-NTCP/SLC10A1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-1958R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:NTCP/SLC10A1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:21-120/349
- Form:Liquid
- Gene ID:6554
- Antigen synonyms:Na(+)/taurocholate transport protein|NTCP|Na(+)/bile acid cotransporter|SLC10A1|Solute carrier family 10 member 1|GIG29|Cell growth-inhibiting gene 29 protein|Sodium/taurocholate cotransporting polypeptide|Sodium/bile acid cotransporter
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:38 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human NTCP
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The hepatic sodium/bile acid uptake system exhibits broad substrate specificity and transports various non-bile acid organic compounds as well. It is strictly dependent on the extracellular presence of sodium.
Type: Primary
Antigen: NTCP/SLC10A1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/349
Host: Rabbit
Isotype: IgG
Reactivity: Human