Anti-HSTF2 Rabbit Polyclonal Antibody
BOSSBS-1409R
: Bioss
- Antibody type:Primary
- Antigen name:HSTF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:401-536/536
- Form:Liquid
- Gene ID:3298
- Antigen synonyms:Heat shock transcription factor 2|Heat shock factor protein 2|HSF2|HSTF2|HSTF 2|HSF 2
- Modification:Unmodified
- Storage buffer:0,01 M TBS(pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:59 kDa
- Storage temperature:Shipped at 4 °C. Store at –20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HSTF2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
DNA-binding protein that specifically binds heat shock promoter elements (HSE) and activates transcription. In higher eukaryotes, HSF is unable to bind to the HSE unless the cells are heat shocked.
Type: Primary
Antigen: HSTF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Public Immunogen Range: 401-536/536
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat