Anti-IFNA14 Rabbit Polyclonal Antibody (ALEXA FLUOR® 555)
Catalog # BOSSBS-15519R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:IFNA14
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:121-189/189
- Form:Liquid
- Gene ID:3448
- Antigen synonyms:MGC125756|LeIFH|IFN14_HUMAN|Interferon alpha-H|Interferon alpha 14|MGC125757|Interferon alpha-14|Interferon lambda2H|IFN-alpha-14|Interferon alpha H|IFNA14|LeIF H|Interferon lambda-2-H
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:20 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human IFNA14
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Type: Primary
Antigen: IFNA14
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 555
Public Immunogen Range: 121-189/189
Host: Rabbit
Isotype: IgG
Reactivity: