SDS Protein Gel Loading Solution, 2X
Catalog # MSPP351-082-661
Supplier: MARKET SOURCE PART PROCESS
CAS Number:
Specifications
- Pk:50 mL
- Description:SDS protein gel loading solution, 2X
- Environmentally Preferable:
- Sterility:Sterile filtered
- Test description:Endotoxin and mycoplasma tested
- Shelf life:36 months
- Shipping temperature:Ambient
Specifications
About this item
SDS Protein Gel Loading Solution, 2X is formulated at the traditional 2X stocks, for standard volume of sample to be loaded per well. Simply load the gel sample wells and visualise progress of the electrophoresis run by monitoring the location of the blue dye front.
- Sterile filtered
- Endotoxin and Performance tested
Lane Marker Sample Buffers may be used in denaturing gels of all kinds and are compatible with Coomassie dye and silver staining techniques.
SDS Protein Gel Loading Solution, 2X is a convenient and ready-to-use solution for SDS-polyacrylamide gel electrophoresis (SDS-PAGE) and Western blotting.
Contents: Tris base, 4% SDS, bromphenol (bromophenol) blue 0,2%, glycerol 20%, and molecular biology grade water.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Ordering information: Optional - add dithiothreitol (0,2ml 1M DTT per ml) or 2-mercaptoethanol (0,02 ml 2-ME per ml) immediately before use.
Caution: Research use only.