Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sunset yellow FCF
Sunset yellow FCF, powder
Sunset yellow FCF, powder
Sunset yellow FCF, powder
Catalog # TCIAF0145-25G
Supplier:
TCI
CAS Number:
Expand image
Sunset yellow FCF, powder
Catalog # TCIAF0145-25G
Supplier:
TCI
CAS Number:
Specifications
Pk:
25 g
Seq:
3308
Specifications
About this item
Product Details
Documentation
:
:
452.36 g/mol
CAS Number:
2783-94-0
Formula:
C₁₆H₁₀N₂Na₂O₇S₂
MDL Number:
MFCD00036437
Product Details
Documentation