Order Entry
Spain
ContactUsLinkComponent
 
Phenol ≥99.0%, fused crystals USP
 
Phenol ≥99.0%, fused crystals USP
 

 

  • Assay (Anhydrous Basis):
    99.0 - 100.5 %
  • Clarity of Solution and Reaction:
    To Pass Test
  • Congealing Temperature:
    39°C min.
  • Water Determination:
    max. 0.5 %
  • Nonvolatile Residue:
    max. 0.05 %

 

 

Frequently Bought Together