Order Entry
Spain
ContactUsLinkComponent
Riboflavine (vitamin B2) ≥98.0% FCC
Riboflavine (vitamin B2) ≥98.0% FCC
Catalog # SPCMR1032-25GMGL
Supplier:  Spectrum Chemical
CAS Number:  
Riboflavine (vitamin B2) ≥98.0% FCC
Catalog # SPCMR1032-25GMGL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Assay (Dried Basis):
    98.0 - 102.0 %
  • Specific Rotation:
    +56.5 to +59.5
  • Loss On Drying:
    max. 1.5 %
  • Lumiflavin:
    To Pass Test
  • Residue On Ignition:
    max. 0.3 %

Specification Test Results

About this item

 :
 :376.37 g/mol
Storage Temperature:Ambient
CAS Number:83-88-5
Formula:C₁₇H₂₀N₄O₆
MDL Number:MFCD00005022