Order Entry
Spain
ContactUsLinkComponent
L-Threonine ≥98.5%, crystals USP
L-Threonine ≥98.5%, crystals USP
Catalog # SPCMT1056-25GM
Supplier:  Spectrum Chemical
CAS Number:  
L-Threonine ≥98.5%, crystals USP
Catalog # SPCMT1056-25GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Assay:
    98.5% Min.

Specification Test Results

About this item

 :
 :119.12 g/mol
Storage Temperature:Ambient
EINECS:200-774-1
CAS Number:72-19-5
Formula:C₄H₉NO₃
MDL Number:MFCD00064270