Order Entry
Spain
ContactUsLinkComponent
L-Threonine ≥98.5%, crystals FCC
L-Threonine ≥98.5%, crystals FCC
Catalog # SPCMT1057-1KG
Supplier:  Spectrum Chemical
CAS Number:  
L-Threonine ≥98.5%, crystals FCC
Catalog # SPCMT1057-1KG
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly pail
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    98.5% Min.

Specification Test Results

About this item

 :
 :119.12 g/mol
Storage Temperature:Ambient
EINECS:200-774-1
CAS Number:72-19-5
Formula:C₄H₉NO₃
MDL Number:MFCD00064270