EDTA disodium salt dihydrate ≥99.0% FCC
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:6381-92-6
:
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037
:372.24 g/mol
Storage Temperature:Ambient
Specification Test Results
- Assay (C₁₀H₁₄O₈N₂Na₂2H₂O):99.0 - 101.0 %
- pH of a 1 in 100 Solution:4.3 - 4.7
- Calcium (Ca):To Pass Test
- Lead (Pb):max. 10 mg/kg
- Nitrilotriacetic Acid[N[(HOCOCH₂)₃N]:To Pass Test
Specification Test Results
Frequently Bought Together