Order Entry
Spain
ContactUsLinkComponent
Sodium chloride, crystal disk for spectroscopy, for CO₂ lasers, polished
Sodium chloride, crystal disk for spectroscopy, for CO₂ lasers, polished
Catalog # INCR0002A-82
CAS Number:  
Sodium chloride, crystal disk for spectroscopy, for CO₂ lasers, polished
Catalog # INCR0002A-82
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :58.44 g/mol
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477