Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Glycogen
Glycogen, powder
Glycogen, powder
Glycogen, powder
SPCMGL160-5GM
:
Spectrum Chemical
:
Glycogen, powder
SPCMGL160-5GM
:
Spectrum Chemical
:
Pack type:
Glass, amber
Pk:
5 g
Seq:
3173
:
:
666.579 g/mol
:
Refrigerator
:
9005-79-2
:
C₂₄H₄₂O₂₁
:
MFCD00081547