Sodium dihydrogen phosphate, anhydrous ≥98.0% USP
Catalog # SPCMSO187-2.5KG
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Poly bottle
- Pk:2,5 kg
- Seq:4076
Specifications
Specification Test Results
- Assay (NaH₂PO₄):98.0 - 103.0 %
- pH of a 5% Solution:4.1 - 4.5
- Water:max. 2.0 %
- Al, Ca, & Related Elements:To Pass Test
- Arsenic (As):max. 8 ppm
Specification Test Results
About this item
:
:119.98 g/mol
Storage Temperature:Ambient
CAS Number:7558-80-7
Formula:NaH₂PO₄
MDL Number:MFCD00003527