Erythrosin B disodium salt
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:16423-68-0
:568-63-8
Formula:C₂₀H₆I₄Na₂O₅
MDL Number:MFCD00144257
:879.86 g/mol
EINECS:240-474-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together