Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Valganciclovir, Reference Standards

 :   USP
Valganciclovir, Reference Standards
 :  USP
 
Valganciclovir, Reference Standards

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

 

 

  • Catalog No:
  • 1708004.
  • 1708015.
  • Description:
  • Valganciclovir Hydrochloride
  • D-Valganciclovir
  • Synonym(s):
  • 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)-methoxy]-3-hydroxypropyl D-valine ester monohydrochloride
  • CAS No.:
  • 175865-59-5

 

Product Family Options

 

  • Description
    Standards
    Pack type
    Pk
     
     
  • Valganciclovir Hydrochloride
    USP reference standard
    500 mg
       
       
    1 * 500 mg
    1 * 500 mg - 628,00 €
    628,00 €
    1 * 500 mg
       628,00 €

     
     
    Specifications
    Cat. No.
    1708004.
    Description
    Valganciclovir Hydrochloride
    Synonym(s)
    CAS No.
    175865-59-5
  • D-Valganciclovir
    USP reference standard
    Vial
    10 mg
       
       
    1 * 10 mg
    1 * 10 mg - 1 240,00 €
    1 240,00 €
    1 * 10 mg
       1 240,00 €

     
     
    Specifications
    Cat. No.
    1708015.
    Description
    D-Valganciclovir
    Synonym(s)
    2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)-methoxy]-3-hydroxypropyl D-valine ester monohydrochloride
    CAS No.