Order Entry
Spain
Orders LinkContactUsLinkComponent
Thimerosal, Reference Standards

Thimerosal, Reference Standards

Supplier:   USP
Thimerosal, Reference Standards
Supplier:  USP
Certificates
Thimerosal, Reference Standards

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1659000.
  • 1659001.
  • Description:
  • Thimerosal
  • Thimerosal Related Compound A
  • Synonym(s):
  • 2,2'-Disulfanediyldibenzoic acid
  • CAS No.:
  • 54-64-8
  • 119-80-2
  • Formula:
  • C₁₄H₁₀O₄S₂
  • UN No.:
  • 2025

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1659000.
    Thimerosal
    Thimerosal
    USP reference standard
    Vial
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 382,00 €
    382,00 €
    1 * 500 mg
       382,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1659000.
    Description
    Thimerosal
    Synonym(s)
    CAS No.
    54-64-8
    Formula
    UN No.
    2025
  • 1659001.
    Thimerosal Related Compound A
    Thimerosal Related Compound A
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 210,00 €
    1 210,00 €
    1 * 50 mg
       1 210,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1659001.
    Description
    Thimerosal Related Compound A
    Synonym(s)
    2,2'-Disulfanediyldibenzoic acid
    CAS No.
    119-80-2
    Formula
    C₁₄H₁₀O₄S₂
    UN No.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.