Nepafenac, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1460317.
- 1460339.
- 1460328.
- Description:
- Nepafenac
- Nepafenac Related Compound B
- Nepafenac Related Compound A
- Synonym(s):
- 7-Benzoylindolin-2-one
- 2-{2-Amino-3-[hydroxy(phenyl)methyl]phenyl}acetamide
- CAS No.:
- 78281-72-8
- 51135-38-7
- 1246956-18-2
- UN No.:
- 2811
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1460317.NepafenacSpecifications:
More
Specifications
Cat. No.1460317.DescriptionNepafenacSynonym(s)CAS No.78281-72-8UN No.2811 - 1460339.Nepafenac Related Compound BNepafenac Related Compound BUSP reference standardVial25 mgLog in for availability1 * 25 mg1 100,00 €Specifications:
More
Specifications
Cat. No.1460339.DescriptionNepafenac Related Compound BSynonym(s)7-Benzoylindolin-2-oneCAS No.51135-38-7UN No. - 1460328.Nepafenac Related Compound ANepafenac Related Compound AUSP reference standardVial25 mgLog in for availability1 * 25 mg1 120,00 €Specifications:
More
Specifications
Cat. No.1460328.DescriptionNepafenac Related Compound ASynonym(s)2-{2-Amino-3-[hydroxy(phenyl)methyl]phenyl}acetamideCAS No.1246956-18-2UN No.



