Order Entry
Spain
Orders LinkContactUsLinkComponent
Levocarnitine, Reference Standards

Levocarnitine, Reference Standards

Supplier:   USP
Levocarnitine, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1359903.
  • 1359925.
  • Description:
  • Levocarnitine
  • Levocarnitine Related Compound A
  • Synonym(s):
  • 2-propen-1-aminium, 3-carboxy-N,N,N-trimethyl-, chloride
  • CAS No.:
  • 541-15-1
  • 6538-82-5
  • Formula:
  • C₇H₁₄ClNO₂

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1359903.
    Levocarnitine
    Levocarnitine
    USP reference standard
    Vial
    400 mg
    Log in for availability
    Log in for availability
    1 * 400 mg
    1 * 400 mg - 400,00 €
    400,00 €
    1 * 400 mg
       400,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1359903.
    Description
    Levocarnitine
    Synonym(s)
    CAS No.
    541-15-1
    Formula
  • 1359925.
    Levocarnitine Related Compound A
    Levocarnitine Related Compound A
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 1 180,00 €
    1 180,00 €
    1 * 100 mg
       1 180,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1359925.
    Description
    Levocarnitine Related Compound A
    Synonym(s)
    2-propen-1-aminium, 3-carboxy-N,N,N-trimethyl-, chloride
    CAS No.
    6538-82-5
    Formula
    C₇H₁₄ClNO₂