Order Entry
Spain
Orders LinkContactUsLinkComponent
Etravirine, Reference Standards
Etravirine, Reference Standards
Supplier:   USP
Etravirine, Reference Standards
Supplier:  USP
Certificates
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1266824.
  • 1266835.
  • Description:
  • Etravirine
  • Etravirine System Suitability Mixture
  • CAS No.:
  • 269055-15-4
Specifications
Product Family Options
Product Information
  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1266824.
    Etravirine
    Etravirine
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 880,00 €
    880,00 €
    1 * 100 mg
       880,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1266824.
    Description
    Etravirine
    CAS No.
    269055-15-4
  • 1266835.
    Etravirine System Suitability Mixture
    Etravirine System Suitability Mixture
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 360,00 €
    1 360,00 €
    1 * 15 mg
       1 360,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1266835.
    Description
    Etravirine System Suitability Mixture
    CAS No.
Recommendations