Order Entry
Spain
Orders LinkContactUsLinkComponent
Collagenase, Reference Standards

Collagenase, Reference Standards

Supplier:   USP
Collagenase, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Caution: Cold Shipment required

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1148089.
  • 1148090.
  • Description:
  • Collagenase I
  • Collagenase II
  • CAS No.:
  • 9001-12-1
  • 9001-12-1

Specifications

Product Family Options

Product Information

  • Standards
    Pk Info
    Pack type
    Pk
    Availability
    Price
  • 1148089.
    Collagenase I
    Collagenase I
    USP reference standard
    2 x 0.5 mL
    Vial
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 423,00 €
    423,00 €
    1 * 1 mL
       423,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1148089.
    Description
    Collagenase I
    CAS No.
    9001-12-1
  • 1148090.
    Collagenase II
    Collagenase II
    USP reference standard
    2 x 0.5 mL
    Vial
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 423,00 €
    423,00 €
    1 * 1 mL
       423,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1148090.
    Description
    Collagenase II
    CAS No.
    9001-12-1