Order Entry
Spain
Orders LinkContactUsLinkComponent
Cidofovir, Reference Standards

Cidofovir, Reference Standards

 :   USP
Cidofovir, Reference Standards
 :  USP
 
Cidofovir, Reference Standards

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

 

 

 

  • Catalog No:
  • 1133853.
  • 1133864.
  • 1133875.
  • 1133886.
  • Description:
  • Cidofovir
  • Cidofovir Enantiomer
  • Cidofovir Related Compound A
  • Cidofovir Related Compound B
  • Synonym(s):
  • 1-[(R)-3-Hydroxy-2-(phosphonomethoxy)propyl]cytosine dihydrate
  • 1-[(S)-3-Hydroxy-2-(O-ethylphosphonomethoxy)propyl]cytosine
  • 1-[(S)-3-Hydroxy-2-(O,O-diethylphosphonomethoxy)propyl]cytosine hydrochloride
  • CAS No.:
  • 149394-66-1
  • 1312776-50-3

 

Product Family Options

 

  • Description
    Standards
    Pack type
    Pk
     
     
  • Cidofovir
    USP reference standard
    Vial
    100 mg
       
       
    1 * 100 mg
    1 * 100 mg - 935,00 €
    935,00 €
    1 * 100 mg
       935,00 €

     
     
    Specifications
    Cat. No.
    1133853.
    Description
    Cidofovir
    Synonym(s)
    CAS No.
    149394-66-1
  • Cidofovir Enantiomer
    USP reference standard
    Vial
    15 mg
       
       
    1 * 15 mg
    1 * 15 mg - 501,00 €
    501,00 €
    1 * 15 mg
       501,00 €

     
     
    Specifications
    Cat. No.
    1133864.
    Description
    Cidofovir Enantiomer
    Synonym(s)
    1-[(R)-3-Hydroxy-2-(phosphonomethoxy)propyl]cytosine dihydrate
    CAS No.
  • Cidofovir Related Compound A
    USP reference standard
    Vial
    15 mg
       
       
    1 * 15 mg
    1 * 15 mg - 785,00 €
    785,00 €
    1 * 15 mg
       785,00 €

     
     
    Specifications
    Cat. No.
    1133875.
    Description
    Cidofovir Related Compound A
    Synonym(s)
    1-[(S)-3-Hydroxy-2-(O-ethylphosphonomethoxy)propyl]cytosine
    CAS No.
    1312776-50-3
  • Cidofovir Related Compound B
    USP reference standard
    Vial
    15 mg
       
       
    1 * 15 mg
    1 * 15 mg - 808,00 €
    808,00 €
    1 * 15 mg
       808,00 €

     
     
    Specifications
    Cat. No.
    1133886.
    Description
    Cidofovir Related Compound B
    Synonym(s)
    1-[(S)-3-Hydroxy-2-(O,O-diethylphosphonomethoxy)propyl]cytosine hydrochloride
    CAS No.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.