Order Entry
Spain
Orders LinkContactUsLinkComponent
Ammonium Phosphate Dibasic, Reference Standard

Ammonium Phosphate Dibasic, Reference Standard

Ammonium Phosphate Dibasic, USP reference standard, Vial, 1 g
Catalog # 1029986.
Supplier:   USP
Ammonium Phosphate Dibasic, Reference Standard
Catalog # 1029986.
Supplier:  USP
CAS Number:  
Ammonium Phosphate Dibasic, Reference Standard

Ammonium Phosphate Dibasic, USP reference standard, Vial, 1 g

CAS Number:  
Price
196,00 €
Log in for availability
1 * 1 g
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    1 g
  • Standards:
    USP reference standard
  • Description:
    Ammonium Phosphate Dibasic
  • Cat. No.:
    1029986.
  • Formula:
    (NH₄)₂HPO₄
  • Description:
    Ammonium Phosphate Dibasic
  • CAS No.:
    7783-28-0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.