Order Entry
Spain
Orders LinkContactUsLinkComponent
Acetohydroxamic Acid, Reference Standard

Acetohydroxamic Acid, Reference Standard

Acetohydroxamic Acid, USP reference standard, 200 mg
Catalog # 1006506.
Supplier:   USP
Acetohydroxamic Acid, Reference Standard
Catalog # 1006506.
Supplier:  USP
CAS Number:  
Acetohydroxamic Acid, Reference Standard

Acetohydroxamic Acid, USP reference standard, 200 mg

CAS Number:  
Price
363,00 €
Log in for availability
1 * 200 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • Description:
    Acetohydroxamic Acid
  • Cat. No.:
    1006506.
  • Formula:
    C₂H₅NO₂
  • Description:
    Acetohydroxamic Acid
  • CAS No.:
    546-88-3

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.